Anti-C15orf40 Mouse Monoclonal Antibody [clone: OTI1D9]

Proveedor: OriGene
ORIGTA501404EA 603 EUR
ORIGTA501404
Anti-C15orf40 Mouse Monoclonal Antibody [clone: OTI1D9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500 Immunofluorescence: 1:100

Type: Primary
Antigen: C15orf40 (chromosome 15 open reading frame 40)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR