Anti-MOB1B Mouse Monoclonal Antibody [clone: OTI3G9]

Proveedor: OriGene
ORIGTA501363EA 603 EUR
ORIGTA501363
Anti-MOB1B Mouse Monoclonal Antibody [clone: OTI3G9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: MOB1B (MOB kinase activator 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR