Anti-C21orf59 Mouse Monoclonal Antibody [clone: OTI2E5]

Proveedor: OriGene
ORIGTA501348EA 603 EUR
ORIGTA501348
Anti-C21orf59 Mouse Monoclonal Antibody [clone: OTI2E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C21orf59 (chromosome 21 open reading frame 59)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR