Anti-TTC32 Mouse Monoclonal Antibody [clone: OTI5C12]

Proveedor: OriGene
ORIGTA501341EA 581 EUR
ORIGTA501341
Anti-TTC32 Mouse Monoclonal Antibody [clone: OTI5C12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: TTC32 (tetratricopeptide repeat domain 32)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR