Anti-TTC32 Mouse Monoclonal Antibody [clone: OTI1F4]

Proveedor: OriGene
ORIGTA501340EA 581 EUR
ORIGTA501340
Anti-TTC32 Mouse Monoclonal Antibody [clone: OTI1F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TTC32 (tetratricopeptide repeat domain 32)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR