Anti-CD5 Mouse Monoclonal Antibody [clone: OTI4H3]

Proveedor: OriGene
ORIGTA501333EA 603 EUR
ORIGTA501333
Anti-CD5 Mouse Monoclonal Antibody [clone: OTI4H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:100; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR