Anti-PANK2 Mouse Monoclonal Antibody [clone: OTI3H9]

Proveedor: OriGene
ORIGTA501321EA 609 EUR
ORIGTA501321
Anti-PANK2 Mouse Monoclonal Antibody [clone: OTI3H9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PANK2 (pantothenate kinase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR