Anti-HIBCH Mouse Monoclonal Antibody [clone: OTI2A5]

Proveedor: OriGene
ORIGTA501318EA 581 EUR
ORIGTA501318
Anti-HIBCH Mouse Monoclonal Antibody [clone: OTI2A5]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH (3-hydroxyisobutyryl-CoA hydrolase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR