Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: OTI4F7]

Proveedor: OriGene
ORIGTA501315EA 603 EUR
ORIGTA501315
Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: OTI4F7]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IGF2BP2 (insulin-like growth factor 2 mRNA binding protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR