Anti-HID1 Mouse Monoclonal Antibody [clone: OTI2F4]

Proveedor: OriGene
ORIGTA501311EA 608 EUR
ORIGTA501311
Anti-HID1 Mouse Monoclonal Antibody [clone: OTI2F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HID1 (HID1 domain containing)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR