Anti-HSPB7 Mouse Monoclonal Antibody [clone: OTI1D11]

Proveedor: OriGene
ORIGTA501310EA 586 EUR
ORIGTA501310
Anti-HSPB7 Mouse Monoclonal Antibody [clone: OTI1D11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HSPB7 (heat shock 27kDa protein family, member 7 (cardiovascular))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR