Anti-USP5 Mouse Monoclonal Antibody [clone: OTI2E3]

Proveedor: OriGene
ORIGTA501290EA 586 EUR
ORIGTA501290
Anti-USP5 Mouse Monoclonal Antibody [clone: OTI2E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50

Type: Primary
Antigen: USP5 (ubiquitin specific peptidase 5 (isopeptidase T))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR