Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: OTI3C8]

Proveedor: OriGene
ORIGTA501274EA 603 EUR
ORIGTA501274
Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: OTI3C8]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IGF2BP2 (insulin-like growth factor 2 mRNA binding protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR