Anti-TACC3 Mouse Monoclonal Antibody [clone: OTI6H4]

Proveedor: OriGene
ORIGTA501264EA 603 EUR
ORIGTA501264
Anti-TACC3 Mouse Monoclonal Antibody [clone: OTI6H4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TACC3 (transforming, acidic coiled-coil containing protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR