Anti-DAPK2 Mouse Monoclonal Antibody [clone: OTI1B10]

Proveedor: OriGene
ORIGTA501204EA 603 EUR
ORIGTA501204
Anti-DAPK2 Mouse Monoclonal Antibody [clone: OTI1B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: DAPK2 (death-associated protein kinase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR