Anti-BSG Mouse Monoclonal Antibody [clone: OTI5D8]

Proveedor: OriGene
ORIGTA501190EA 603 EUR
ORIGTA501190
Anti-BSG Mouse Monoclonal Antibody [clone: OTI5D8]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR