Anti-NME4 Mouse Monoclonal Antibody [clone: OTI2B10]
Proveedor: OriGene
ORIGTA501177EA 603 EUR
ORIGTA501177
Anti-NME4 Mouse Monoclonal Antibody [clone: OTI2B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: NME4 (NME/NM23 nucleoside diphosphate kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human