Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI1E12]

Proveedor: OriGene
ORIGTA501172EA 603 EUR
ORIGTA501172
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: OTI1E12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2 (cytochrome P450, family 1, subfamily A, polypeptide 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR