Anti-CDH13 Mouse Monoclonal Antibody [clone: OTI3H6]
Proveedor: OriGene
ORIGTA501158EA 608 EUR
ORIGTA501158
Anti-CDH13 Mouse Monoclonal Antibody [clone: OTI3H6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: CDH13 (cadherin 13)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human