Anti-ARNT Mouse Monoclonal Antibody [clone: OTI1D10]

Proveedor: OriGene
ORIGTA501148EA 603 EUR
ORIGTA501148
Anti-ARNT Mouse Monoclonal Antibody [clone: OTI1D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ARNT (aryl hydrocarbon receptor nuclear translocator)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR