Anti-PRKAR2A Mouse Monoclonal Antibody [clone: OTI1F8]

Proveedor: OriGene
ORIGTA501146EA 603 EUR
ORIGTA501146
Anti-PRKAR2A Mouse Monoclonal Antibody [clone: OTI1F8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKAR2A (protein kinase, cAMP-dependent, regulatory, type II, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR