Anti-PRKAR2A Mouse Monoclonal Antibody [clone: OTI1A7]
Proveedor: OriGene
ORIGTA501129EA 603 EUR
ORIGTA501129
Anti-PRKAR2A Mouse Monoclonal Antibody [clone: OTI1A7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: PRKAR2A (protein kinase, cAMP-dependent, regulatory, type II, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human