Anti-CAPZA1 Mouse Monoclonal Antibody [clone: OTI7C8]

Proveedor: OriGene
ORIGTA501125EA 608 EUR
ORIGTA501125
Anti-CAPZA1 Mouse Monoclonal Antibody [clone: OTI7C8]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:200-500

Type: Primary
Antigen: CAPZA1 (capping protein (actin filament) muscle Z-line, alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR