Anti-BTK Mouse Monoclonal Antibody [clone: OTI3E9]

Proveedor: OriGene
ORIGTA501100EA 603 EUR
ORIGTA501100
Anti-BTK Mouse Monoclonal Antibody [clone: OTI3E9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: BTK (Bruton agammaglobulinemia tyrosine kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR