Anti-PIM2 Mouse Monoclonal Antibody [clone: OTI8G10]

Proveedor: OriGene
ORIGTA501060EA 581 EUR
ORIGTA501060
Anti-PIM2 Mouse Monoclonal Antibody [clone: OTI8G10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PIM2 (pim-2 oncogene)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR