Anti-LIPG Mouse Monoclonal Antibody [clone: OTI3C4]

Proveedor: OriGene
ORIGTA501059EA 603 EUR
ORIGTA501059
Anti-LIPG Mouse Monoclonal Antibody [clone: OTI3C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: LIPG (lipase, endothelial)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR