Anti-TUBA8 Mouse Monoclonal Antibody [clone: OTI2E9]

Proveedor: OriGene
ORIGTA501038EA 581 EUR
ORIGTA501038
Anti-TUBA8 Mouse Monoclonal Antibody [clone: OTI2E9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100;

Type: Primary
Antigen: TUBA8 (tubulin, alpha 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR