Anti-MCL1 Mouse Monoclonal Antibody [clone: OTI10F6]

Proveedor: OriGene
ORIGTA501036EA 603 EUR
ORIGTA501036
Anti-MCL1 Mouse Monoclonal Antibody [clone: OTI10F6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: MCL1 (myeloid cell leukemia 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR