Anti-KCTD5 Mouse Monoclonal Antibody [clone: OTI3C8]

Proveedor: OriGene
ORIGTA501035EA 628 EUR
ORIGTA501035
Anti-KCTD5 Mouse Monoclonal Antibody [clone: OTI3C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: KCTD5 (potassium channel tetramerization domain containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR