Anti-SIL1 Mouse Monoclonal Antibody [clone: OTI3B11]

Proveedor: OriGene
ORIGTA501010EA 608 EUR
ORIGTA501010
Anti-SIL1 Mouse Monoclonal Antibody [clone: OTI3B11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SIL1 (SIL1 nucleotide exchange factor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR