Anti-APP Mouse Monoclonal Antibody [clone: OTI4C3]

Proveedor: OriGene
ORIGTA501006EA 603 EUR
ORIGTA501006
Anti-APP Mouse Monoclonal Antibody [clone: OTI4C3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR