Anti-SIL1 Mouse Monoclonal Antibody [clone: OTI1G7]

Proveedor: OriGene
ORIGTA500996EA 603 EUR
ORIGTA500996
Anti-SIL1 Mouse Monoclonal Antibody [clone: OTI1G7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SIL1 (SIL1 nucleotide exchange factor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR