Anti-MCL1 Mouse Monoclonal Antibody [clone: OTI2E11]

Proveedor: OriGene
ORIGTA500986EA 608 EUR
ORIGTA500986
Anti-MCL1 Mouse Monoclonal Antibody [clone: OTI2E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: MCL1 (myeloid cell leukemia 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR