Anti-SRR Mouse Monoclonal Antibody [clone: OTI5G3]

Proveedor: OriGene
ORIGTA500981EA 603 EUR
ORIGTA500981
Anti-SRR Mouse Monoclonal Antibody [clone: OTI5G3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SRR (serine racemase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR