Anti-FAHD2A Mouse Monoclonal Antibody [clone: OTI5G1]
Proveedor: OriGene
ORIGTA500979EA 581 EUR
ORIGTA500979
Anti-FAHD2A Mouse Monoclonal Antibody [clone: OTI5G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: FAHD2A (fumarylacetoacetate hydrolase domain containing 2A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human