Anti-BHMT Mouse Monoclonal Antibody [clone: OTI10B3]

Proveedor: OriGene
ORIGTA500960EA 603 EUR
ORIGTA500960
Anti-BHMT Mouse Monoclonal Antibody [clone: OTI10B3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR