Anti-IFIT1 Mouse Monoclonal Antibody [clone: OTI3G8]

Proveedor: OriGene
ORIGTA500948EA 609 EUR
ORIGTA500948
Anti-IFIT1 Mouse Monoclonal Antibody [clone: OTI3G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: IFIT1 (interferon-induced protein with tetratricopeptide repeats 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR