Anti-PLEK Mouse Monoclonal Antibody [clone: OTI6E7]

Proveedor: OriGene
TA500902
ORIGTA500902EA 593 EUR
ORIGTA500902
Anti-PLEK Mouse Monoclonal Antibody [clone: OTI6E7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PLEK (pleckstrin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR