Anti-TF Mouse Monoclonal Antibody [clone: OTI5G2]

Proveedor: OriGene
ORIGTA500848EA 608 EUR
ORIGTA500848
Anti-TF Mouse Monoclonal Antibody [clone: OTI5G2]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TF (transferrin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR