Anti-ATP5B Mouse Monoclonal Antibody [clone: OTI6C10]

Proveedor: OriGene
ORIGTA500838EA 603 EUR
ORIGTA500838
Anti-ATP5B Mouse Monoclonal Antibody [clone: OTI6C10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ATP5B (ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR