Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI1D11]

Proveedor: OriGene
ORIGTA500829EA 608 EUR
ORIGTA500829
Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI1D11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: GBE1 (glucan (1,4-alpha-), branching enzyme 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR