Anti-HSPA1A Mouse Monoclonal Antibody [clone: OTI1B2]

Proveedor: OriGene
ORIGTA500827EA 603 EUR
ORIGTA500827
Anti-HSPA1A Mouse Monoclonal Antibody [clone: OTI1B2]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: HSPA1A (heat shock 70kDa protein 1A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR