Anti-PRL Mouse Monoclonal Antibody [clone: OTI2C3]

Proveedor: OriGene
ORIGTA500818EA 581 EUR
ORIGTA500818
Anti-PRL Mouse Monoclonal Antibody [clone: OTI2C3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PRL (prolactin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR