Anti-HSD17B10 Mouse Monoclonal Antibody [clone: OTI5E2]

Proveedor: OriGene
ORIGTA500813EA 608 EUR
ORIGTA500813
Anti-HSD17B10 Mouse Monoclonal Antibody [clone: OTI5E2]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: HSD17B10 (hydroxysteroid (17-beta) dehydrogenase 10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR