Anti-RPA2 Mouse Monoclonal Antibody [clone: OTI7F4]

Proveedor: OriGene
ORIGTA500812EA 603 EUR
ORIGTA500812
Anti-RPA2 Mouse Monoclonal Antibody [clone: OTI7F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: RPA2 (replication protein A2, 32kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR