Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI2H10]
Proveedor: OriGene
ORIGTA500805EA 581 EUR
ORIGTA500805
Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI2H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100
Type: Primary
Antigen: TTLL12 (tubulin tyrosine ligase-like family, member 12)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human