Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI4E6]

Proveedor: OriGene
ORIGTA500779EA 603 EUR
ORIGTA500779
Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI4E6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: GBE1 (glucan (1,4-alpha-), branching enzyme 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR