Anti-PPME1 Mouse Monoclonal Antibody [clone: OTI2C5]

Proveedor: OriGene
ORIGTA500768EA 581 EUR
ORIGTA500768
Anti-PPME1 Mouse Monoclonal Antibody [clone: OTI2C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PPME1 (protein phosphatase methylesterase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR