Anti-AKR1A1 Mouse Monoclonal Antibody [clone: OTI3C7]

Proveedor: OriGene
ORIGTA500767EA 603 EUR
ORIGTA500767
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: OTI3C7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: AKR1A1 (aldo-keto reductase family 1, member A1 (aldehyde reductase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR