Anti-HDAC10 Mouse Monoclonal Antibody [clone: OTI4D4]

Proveedor: OriGene
ORIGTA500752EA 581 EUR
ORIGTA500752
Anti-HDAC10 Mouse Monoclonal Antibody [clone: OTI4D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: HDAC10 (histone deacetylase 10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR